Learn More
Aviva Systems Biology Corp FGF13 Antibody - middle region (ARP55412_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55412_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FGF13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FGF13. Alternate names: FGF2, FHF2, FHF-2, FGF-13ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61497