Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549426

Catalog No. PA549426

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Probably involved in nervous system development and function.
Specifications

Specifications

FGF14
Polyclonal
PBS with 0.09% sodium azide
Q92915, P70379, Q8R5L7
Fibroblast growth factor 14, FGF-14, Fibroblast growth factor homologous factor 4, FHF-4, FGF14, FHF4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
14169, 2259, 63851
Western Blot
Unconjugated
FGF14
Liquid
FGF14
KLH conjugated synthetic peptide between 207-241 amino acids from the C-terminal region of human FGF14.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit