Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF16 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548297
Description
Predicted to react with rat based on sequence homology.
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation, and is required for normal cardiomyocyte proliferation and heart development.
Specifications
FGF16 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9ESL8 | |
FGF16 | |
KLH conjugated synthetic peptide between 22-50 amino acids from the N-terminal region of human FGF16. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Fgf16 | |
Fibroblast growth factor 16, FGF-16, FGF16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80903 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction