Learn More
FGF19 293T Cell Overexpression Lysate 2 (Denatured), Abnova
Manufacturer:  Abnova Corporation H00009965T02
Catalog No. 89-001-854
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
NM_005117
6.8
Transient overexpression cell Lysate was tested with Anti-FGF19 antibody (H00009965-B02) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
FGF19 293T Cell Transient Overexpression Lysate(Denatured)
23.87
pCMV-FGF19 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.