Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FGF21 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518811
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.
Specifications
FGF21 | |
Polyclonal | |
Unconjugated | |
FGF21 | |
FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196 | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
26291 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9NSA1 | |
FGF21 | |
Synthetic peptide sequence (CQRPDGALYGSLH) corresponding to the internal amino acids of FGF21 (aa 103-116). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction