Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551079

Catalog No. PA551079


Add to cart

Description

Description

FGF6 Polyclonal Antibody detects endogenous levels of FGF6 protein.

Plays an important role in the regulation of cell proliferation, cell differentiation, angiogenesis and myogenesis, and is required for normal muscle regeneration. {ECO:0000269; PubMed:8663044}.
Specifications

Specifications

FGF6
Polyclonal
Unconjugated
FGF6
Liquid
FGF6
Synthesized peptide derived from the C-terminal region of human FGF-6.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P10767, P21658
FGF6, HST2, HSTF2, Fibroblast growth factor 6, FGF-6, Heparin secretory-transforming protein 2, HST-2, HSTF-2, Heparin-binding growth factor 6, HBGF-6
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
14177, 170700, 2251
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit