Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551114

Catalog No. PA551114


Add to cart

Description

Description

FGF8 Polyclonal Antibody detects endogenous levels of FGF8 protein.

Plays an important role in the regulation of embryonic development, cell proliferation, cell differentiation and cell migration. Required for normal brain, eye, ear and limb development during embryogenesis. Required for normal development of the gonadotropin-releasing hormone (GnRH) neuronal system (PubMed:16384934, PubMed:16597617, PubMed:8663044). Plays a role in neurite outgrowth in hippocampal cells (PubMed:21576111). {ECO:0000269; PubMed:16384934, ECO:0000269; PubMed:16597617, ECO:0000269; PubMed:21576111, ECO:0000269; PubMed:8663044}.
Specifications

Specifications

FGF8
Polyclonal
Unconjugated
FGF8
Liquid
FGF8
Synthesized peptide derived from the Internal region of human FGF-8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P55075, P37237
FGF8, AIGF, Fibroblast growth factor 8, FGF-8, Androgen-induced growth factor, AIGF, Heparin-binding growth factor 8, HBGF-8
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
14179, 2253, 29349
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit