Learn More
FGFBP1 Mouse anti-Human, Clone: OTI5F11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808313
Catalog No. 50-166-8768
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FGFBP1 encodes a secreted fibroblast growth factor carrier protein. The encoded protein plays a critical role in cell proliferation, differentiation and migration by binding to fibroblast growth factors and potentiating their biological effects on target cells. The encoded protein may also play a role in tumor growth as an angiogenic switch molecule, and expression of this gene has been associated with several types of cancer including pancreatic and colorectal adenocarcinoma. A pseudogene of this gene is also located on the short arm of chromosome 4.
Specifications
Specifications
FGFBP1
Monoclonal
Unconjugated
FGFBP1
Lyophilized
FGFBP1
Human recombit protein fragment corresponding to amino acids 24-234 of human FGFBP1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI5F11
PBS with 8% trehalose and no preservative; pH 7.3
Q14512
FGF-BP, FGF-BP1, FGFBP, FGFBP-1, HBP17
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
9982