Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FGFBP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558686
Description
Immunogen sequence: PVLRPSVCRE AGPQAHMQQV TSSLKGSPEP NQQPEAGTPS LRPKATVKLT EATQLGKDSM EELGKAKPTT R Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 30%.
This gene encodes a member of the fibroblast growth factor binding protein family. The encoded protein is a serum protein that is selectively secreted by cytotoxic lymphocytes and may be involved in cytotoxic lymphocyte-mediated immunity. An increase in the amount of gene product may be associated with atopic asthma and mild extrinsic asthma.
Specifications
FGFBP2 | |
Polyclonal | |
Unconjugated | |
FGFBP2 | |
37 kDa killer-specific secretory protein; FGF-binding protein 2; FGFBP2; FGF-BP2; FGFBP-2; fibroblast growth factor binding protein 2; fibroblast growth factor-binding protein 2; HBp17-related protein; HBP17RP; HBp17-RP; killer-specific secretory protein of 37 kDa; KSP37; UNQ425/PRO1065 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
83888 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.30 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BYJ0 | |
FGFBP2 | |
Recombinant protein corresponding to Human FGFBP2. Recombinant protein control fragment (Product #RP-96652). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction