Learn More
FGFBP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 247251AP
Catalog No. 50-173-4508
Add to cart
Description
Description
FGFBP3 is a heparin-binding protein which binds to FGF2, prevents binding of FGF2 to heparin and probably inhibits immobilization of FGF2 on extracellular matrix glycosaminoglycans, allowing its release and subsequent activation of FGFR signaling which leads to increased vascular permeability.
Specifications
Specifications
FGFBP3
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q1HCM0, Q8TAT2
C10orf13, FGF binding protein 3, FGF BP3, FGFBP 3, FGFBP3
Rabbit
IgG
150 μL
-20°C
Primary
143282, 72514
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FGFBP3
Liquid
FGFBP3
FGFBP3 Fusion Protein Ag20573
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse