Learn More
FGFR1 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0277-6F10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212308AF532
Catalog No. NP212308A32
Add to cart
Description
Description
FGFR1 Monoclonal antibody specifically detects FGFR1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
FGFR1
MM0277-6F10
Western Blot
basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, CD331, CD331 antigen, CEK, EC 2.7.10, EC 2.7.10.1, FGFBR, FGFR-1, fibroblast growth factor receptor 1, FLGH3, FLJ99988, FLT-2, FLT2H4, Fms-like tyrosine kinase 2, fms-related tyrosine kinase 2, H2, H5, HBGFR, heparin-binding growth factor receptor, hydroxyaryl-protein kinase, KAL 2, KAL2, N-SAM, OGD, Proto-oncogene c-Fgr, soluble FGFR1 variant 1, soluble FGFR1 variant 2
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
FGFR1
The extracellular domain of the Human FGFR1 protein.
0.1 mL
Primary
2260