Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ FGFR4 Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2796355PLATES

 View more versions of this product

Catalog No. NB010310


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Rat FGFR4.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
23.44 pg/mL
2264
RUO
FGFR4
Angiogenesis, Protein Kinase
Rat
FGFR4
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.4 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1500 pg/mL is recommended.
HRP
This solid phase sandwich ELISA utilizes an antibody specific for Rat FGFR4.
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of rabbit anti-FGFR4 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.5 mg/mL of rabbit anti-Rat FGFR4 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 40 ng of recombinant Rat FGFR4.
  • Storage is content dependent.
    Rabbit
    5 Plates
    23.44-1500 pg/mL
    CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase
    Conjugated
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit