Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR4 Rabbit anti-Macaque, Alexa Fluor 700, Clone: 148, Novus Biologicals™

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP290949AF700

Catalog No. NB080291


Add to Cart

Description

Description

FGFR4 Monoclonal specifically detects FGFR4 in Rhesus Macaque samples. It is validated for ELISA.
Specifications

Specifications

FGFR4
Monoclonal
Alexa Fluor 700
CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase
This antibody was obtained from a rabbit immunized with purified, recombinant Rhesus FGFR4 (XP_001087243.1; Met1-Asp369).
0.1 mL
Angiogenesis, Protein Kinase
2264
Store at 4°C in the dark.
IgG
ELISA
148
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Macaque
Purified
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit