Learn More
FGFR5/FGFRL1 Rat anti-Mouse, Alexa Fluor™ 350, Clone: RM0232-7L43, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212294AF350
Catalog No. NP212294A30
Add to cart
Description
Description
FGFR5/FGFRL1 Monoclonal antibody specifically detects FGFR5/FGFRL1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
FGFR5/FGFRL1
RM0232-7L43
Western Blot
FGF homologous factor receptor, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1
Rat
IgG2b
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
FGFRL1
Mouse FGFRL1
0.1 mL
Primary
53834