Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Fggy Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569516
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
FGGY gene ontology annotations related to this gene include neuron cellular homeostasis; pentose metabolic process.
Specifications
Fggy | |
Polyclonal | |
Unconjugated | |
FGGY | |
2310009E04Rik; Fggy; FGGY carbohydrate kinase domain containing; FGGY carbohydrate kinase domain-containing protein; zgc:85818 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
75578 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A2AJL3 | |
FGGY | |
synthetic peptide directed towards the middle region of human Fggy (aa 128-177). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction