Learn More
Abnova™ FGL2 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00010875T01
Catalog No. 89-944-552
Add to cart
Specifications
Specifications
Western Blot
293T
NM_006682.2
pCMV-FGL2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
FGL2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 48.4kDa
Transient overexpression cell lysate was tested with anti-FGL2 antibody (Mfr. No. H00010875-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)