Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FGR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581997
Description
Immunogen sequence: VFCKKLEPVA TAKEDAGLEG DFRSYGAADH YGPDPTKARP ASSFAHIPNY SNFSSQAINP GFLDSGTIRG VSGIGVTLFI ALYDYEARTE DDLTFTKGEK FHILNNTEGD WWEARSLSSG KTGCIPSNYV APVDSIQAEE WYFGKIG.
FGR is a SRC-related kinase present in hematopoietic cells. FGR signals within multiple pathways including within the integrin pathway and downstream of chemokine and cytokine receptors. This protein contains N-terminal sites for myristylation and palmitoylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. It localizes to plasma membrane ruffles, and functions as a negative regulator of cell migration and adhesion triggered by the beta-2 integrin signal transduction pathway. Infection with Epstein-Barr virus results in the overexpression of this protein.
Specifications
FGR | |
Polyclonal | |
Unconjugated | |
FGR | |
c-fgr; c-fgr protooncogene; c-src2; c-src-2 proto-oncogene; EC 2.7.10.2; feline Gardner-Rasheed sarcoma viral oncogene; feline Gardner-Rasheed sarcoma viral oncogene homolog; FGR; FGR protein; FGR proto-oncogene, Src family tyrosine kinase; FLJ43153; Gardner-Rasheed feline sarcoma viral (Fgr) oncogene homolog; Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog; MGC75096; OTTHUMP00000003603; OTTHUMP00000003604; p55c-fgr; p55-c-fgr protein; p55-Fgr; p58c-Fgr; p58-Fgr; proto-oncogene c-Fgr; proto-oncogene tyrosine-protein kinase FGR; SRC2; Tyrosine-protein kinase Fgr; v-fgr feline Gardner-Rasheed sarcoma viral oncogene homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2268 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09769 | |
FGR | |
Recombinant protein corresponding to Human FGR. Recombinant protein control fragment (Product #RP-95911). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction