Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Scientific™ FH10, FH15 and FH30 Peristaltic Tubing Pumps

Peristaltic pumps - enhanced and versatile performance in an ultra-compact, low-maintenance design

 View more versions of this product

Catalog No. 13-310-651


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.015 to 0.08mL/min.
Uses microbore auto-analysis tubing size 0.19mm to 2.79mm I.D.
IP22
50/60Hz
ETL, cETL, CE, RoHS
11.4cm
16.5cm
13.6cm
Start / Stop
UL, cUL (Power Supply) CE, RoHS, ISO9001:2008
10rpm
2 bar
1
Green LED, On/Off
Powder-Coated steel
4.5 in.
6.5 in.
5.3 in.
FH10 Peristaltic Pump System
1
6.5 x 5.3 x 4.5 in. (16.5 x 13.6 x 11.4cm)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit