Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FHIT Monoclonal Antibody (OTI1D7)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527467

Catalog No. PIMA527467


Only null left
Add to Cart

Description

Description

FHIT Monoclonal Antibody for Western Blot, IHC (P)

Alterations and deletions of the FHIT gene are strongly linked to the genesis and establishment of human tumors of the lung, cervix, breast, colon, stomach and pancreas. In normal cells, FHIT may act as a tumor suppressor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FHIT
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P49789
FHIT
Full length human recombinant protein of FHIT produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
OTI1D7
Unconjugated
FHIT
AP3A hydrolase; AP3Aase; AW045638; bis(5'-adenosyl)-triphosphatase; bis5'-adenosyl-triphosphatase; Diadenosine 5',5'''-P1,P3-triphosphate hydrolase; dinucleosidetriphosphatase; FHIT; Fra14A2; FRA3B; fragile histidine triad; fragile histidine triad gene; fragile histidine triad protein
Mouse
Affinity Chromatography
RUO
2272
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.