Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FHL5 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550300

Catalog No. PA550300


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total FHL5 protein.

The protein encoded by this gene is coordinately expressed with activator of cAMP-responsive element modulator (CREM). It is associated with CREM and confers a powerful transcriptional activation function. CREM acts as a transcription factor essential for the differentiation of spermatids into mature spermatozoa. There are multiple polyadenylation sites found in this gene. Alternative splicing results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FHL5
Polyclonal
Unconjugated
FHL5
1700027G07Rik; Act; activator of cAMP-responsive element modulator (CREM) in testis; activator of cAMP-responsive element modulator in testis; Activator of CREM in testis; dJ393D12.2; FHL5; FHL-5; four and a half LIM domains 5; four and a half LIM domains protein 5; LIM protein ACT; LIM-only protein ACT; RP3-393D12.2
Rabbit
Antigen Affinity Chromatography
RUO
9457
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.9 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q5TD97
FHL5
Fusion protein corresponding to a region derived from internal residues of human four and a half LIM domains 5.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.