Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FHL5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen FHL5
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096041
View Documents
Novus Biologicals
NBP304629100UL
100 μg
Each for $320.71
Add to cart
 
NB096042
View Documents
Novus Biologicals
NBP30462920UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

FHL5 Polyclonal antibody specifically detects FHL5 in Human samples. It is validated for Western Blot
Specifications

Specifications

FHL5
Unconjugated
Rabbit
Human
activator of cAMP-responsive element modulator (CREM) in testis, Activator of cAMP-responsive element modulator in testis, Activator of CREM in testis, ACTKIAA0776, dJ393D12.2, FHL-5, FLJ33049, four and a half LIM domains 5,1700027G07Rik, four and a half LIM domains protein 5, LIM protein ACT, RP3-393D12.2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-284 of human FHL5 (NP_065228.4).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
9457
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit