Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FHOD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554524
Description
Immunogen sequence: GVDTELLVYA MTLVNKTLSG LPDQDTFYDV VDCLEELGIA AVSQRHLNKK GTDLDLVEQL NIYEVALRHE DGDETTEPPP SGCRDRRRAS VCSSGGGEHR GLDRRRSRRH SVQSIKSTLS APTSPCSQSA PSFKPNQV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
FHOD3 is a novel member of the formin homology family playing an important role in regulation of the actin cytoskeleton. It contains a DAD (diaphanous auto regulatory) domain, an FH1 (formin homology 1) domain, an FH2 (formin homology 2) domain and a GBD/FH3 (Rho GTPase-binding/formin homology 3) domain. FHOD3 is a cytoplasmic protein where the main part of the protein localizes to actin fibers and the remaining part displays filamentous staining. A 1422 amino acid containing formin-related actin-organizing protein, FHOD3 may cause stress fiber formation together with cell elongation and may associate with nestin/NES-based interfilament (IF). Expression of FHOD3 is seen mainly in heart, kidney, and brain.
Specifications
FHOD3 | |
Polyclonal | |
Unconjugated | |
FHOD3 | |
A930009H06Rik; FH1/FH2 domain-containing protein 3; Fhod3; Fhos2; Formactin2; Formactin-2; Formin homolog overexpressed in spleen 2; formin homology 2 domain containing 3; formin-family protein FHOS2; hFHOS2; KIAA1695; mFHOS2; mKIAA1695 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80206 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2V2M9 | |
FHOD3 | |
Recombinant protein corresponding to Human FHOD3. Recombinant protein control fragment (Product #RP-88593). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction