Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Fiberboard Storage Boxes (Colored) Available on GSA/VA Contract for Federal Government customers only.

Store a variety of vials and tubes with Thermo Scientific™, water-repellant, fiberboard storage boxes, for use in ultra-low temperature or cryogenic freezers.

Manufacturer:  Thermo Scientific™ 1950880

 View more versions of this product

Catalog No. 19-508-80


Add to cart

Description

Description

Store a variety of vials and tubes with Thermo Scientific™, water-repellant, fiberboard storage boxes, for use in ultra-low temperature or cryogenic freezers. Available in packs of 12; choose from yellow, pink, red, blue, green or multi-colored. Grid dividers sold separately.

  • Sturdy, water-repellant fiberboard boxes with covers for sample storage
  • Available in 2  in. high (5cm)
Ordering Information:
Available in packs of 12: choose from yellow, pink, red, blue, green or multi-colored. Grid dividers sold separately.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Available in packs of 12: choose from yellow, pink, red, blue, green or multi-colored. Grid dividers sold separately.

Specifications

Specifications

2 in. (50mm) Water-repellant fiberboard box
Red
Cryo Storage Box
2 in.
5 in.
5 in.
Does not include dividers
Square
Ultra-Low Temperature Freezer
Fiberboard
5 cm
12.7 cm
12.7 cm
5 x 5 x 2 in. (12.7 x 12.7 x 5 cm)
12/Pk., 12/Cs.
0.5 kg
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit