Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Fiberglass Danger Sign: COMBUSTIBLE LIQUIDS

Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

$42.00 - $53.30

Specifications

Color Red on White
Shape Rectangular
Displayed Text COMBUSTIBLE LIQUIDS
Temperature Range (English) -40°F to 194°F
Finish Not Laminated
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-106-222
View Documents
Brady™
131809
10 x 7 in.
Each for $42.00
Add to Cart
 
19-106-223
View Documents
Brady™
131810
14 x 10 in.
Each for $53.30
Add to Cart
 
Description

Description

  • Fiberglass material is abrasion-resistant and chemical-resistant
  • Fiberglass out-performs all other sign materials in terms of strength, durability, and longevity
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
  • Fiberglass out-performs all other sign materials in terms of strength, durability, and longevity
Specifications

Specifications

Red on White
COMBUSTIBLE LIQUIDS
Not Laminated
1 Ea.
Corner holes
-40°F
Rectangular
-40°F to 194°F
Fiberglass
Non-adhesive
194°F
0.0984 in.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit