Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Fiberglass Sign: HIGH VOLTAGE. KEEP AWAY. CONTACT WILL RESULT IN SERIOUS SHOCK OR DEATH
Improve workplace safety with proper signage and markings. This Brady™ fiberglass sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Specifications
Material | Fiberglass |
---|---|
Shape | Rectangular |
Displayed Text | HIGH VOLTAGE. KEEP AWAY. CONTACT WILL RESULT IN SERIOUS SHOCK OR DEATH |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-109-583
|
Brady™
144414 |
14 x 10 in. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
19-109-584
|
Brady™
144415 |
17 x 7 in. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Fiberglass material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Fiberglass out-performs all other sign materials in terms of strength, durability, and longevity
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Fiberglass | |
Rectangular | |
HIGH VOLTAGE. KEEP AWAY. CONTACT WILL RESULT IN SERIOUS SHOCK OR DEATH |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title