Learn More
Thermo Scientific™ Fiberlite™ F10-6 x 500y Fixed-Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Reduce run time with the Thermo Scientific? Fiberlite? F10-6 x 500y Fixed-Angle Rotor, which provides six-place capacity.
Manufacturer:  Thermo Scientific™ 096062114
 View more versions of this product
Catalog No. 09-606-2114
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Reduce run time with the Thermo Scientific™ Fiberlite™ F10-6 x 500y Fixed-Angle Rotor, which provides six-place capacity.
Features:
  • Reduce run-times with higher G-forces achieved through lightweight carbon fiber rotors
  • Corrosion-and fatigue-resistant design for structural integrity
  • Backed by a 15-year warranty
  • Compatible with Thermo Scientific™ Nalgene™ bottles and tubes for complete centrifuge solutions
Compatible with:
Thermo Scientific™ Sorvall™ Evolution™ RC Series, Hitachi™ Himac CR21/CR22 Series, Beckman™ J2, Avanti Series (Except the Avanti J-HC), Thermo Scientific Jouan KR22i Centrifuge.
Recommended for:
Processing of tissue homogenates and other large particles; clarification of lysates, bacterial cell culture harvesting and debris from water studies.
Note:
†Performance may vary by centrifuge model
Description & Specifications
Specifications
23°
2.75 in.
Carbon fiber composite
Beckman™ J2, Avanti Series centrifuges
17,700 x g
160mm
12.7kg
1.6 in.
41mm
Fiberlite F10-6 x 500y Fixed Angle Rotor
6 x 500mL
70mm
10,000rpm
Fixed angle rotor
6.3 in.
28 lb.
6.22 in.
158mm
4592 x g
3413