Learn More
Cole-Parmer Two-Way Articulating Tip Fiberscope, 40"L, 0.236" Dia, 120 Degree  DFS Item
Manufacturer:  Cole-Parmer 0974416
These fiberscopes feature a control knob that allows the viewing tip to flex in directions needed for inspection. Internal fiber optic bundles provide sharp, high-resolution images of limited access areas. The small diameter shaft allows for inspection of internal components — eliminating or reducing equipment tear-down. Fiberscopes feature a shaft that keeps shape when bent or positioned during inspection.
Catalog No. 50-632-511