Learn More
Abnova™ FIBIN 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Supplier:  Abnova™ H00387758T01
Catalog No. 89-944-577
Add to Cart
Description
Description
Specifications
Specifications
293T
FIBIN 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 24.3kDa
Transient overexpression cell lysate was tested with anti-FIBIN antibody (Mfr. No. H00387758-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
NM_203371
pCMV-FIBIN full-length
100 μL
Human
Product Suggestions
Product Suggestions