Learn More
WASP Project Fibral Plate (8 mm) for Delta WASP Printers
SCI_ED
Magnetic flexible build plate for the WASP 3MT. Vastily improves the adhesion to the plate, and the ease of use to remove 3D printed parts
Manufacturer:  WASP Project WU7070
Catalog No. S81229
Add to cart
Description
Description
  • 8 mm fibral aluminium printing bed allows users to obtain a more stable level in the whole printing surface, thanks to its adjustment process.
  • High aluminium thermal conductivity gives improved heating of the bed.
  • This to avoid surfaces adulteration and to quickly release heat to the object and improve the layer adhesion.
Specifications
Specifications
Delta WASP Printers
STEM Project