Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrillin 2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303631100UL
Description
Fibrillin 2 Polyclonal specifically detects Fibrillin 2 in Human, Mouse samples. It is validated for Western Blot.Specifications
Fibrillin 2 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Unconjugated | |
CCA, congenital contractural arachnodactyly, DA9, fibrillin 2, fibrillin 5, fibrillin-2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 2733-2912 of human Fibrillin 2 (NP_001990.2). | |
100 μL | |
Cell Biology, Cellular Markers | |
2201 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction