Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrillin 2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Fibrillin 2
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094812
View Documents
Novus Biologicals
NBP303631100UL
100 μg Each for $320.71
Add to cart
 
NB094813
View Documents
Novus Biologicals
NBP30363120UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Fibrillin 2 Polyclonal antibody specifically detects Fibrillin 2 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

Fibrillin 2
Unconjugated
Rabbit
Cell Biology, Cellular Markers
PBS with 50% glycerol, pH7.3.
2201
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human, Mouse
CCA, congenital contractural arachnodactyly, DA9, fibrillin 2, fibrillin 5, fibrillin-2
Recombinant fusion protein containing a sequence corresponding to amino acids 2733-2912 of human Fibrillin 2 (NP_001990.2).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit