Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrinogen alpha chain Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 206451AP

Catalog No. 50-173-3010


Add to cart

Description

Description

Fibrinogen is a soluble protein found in blood plasma and is produced by the liver cells. The protein is a hexamer consisting of two sets of disulfide linked chains. Fibrinogen is essential for blood coagulation as the precursor of fibrin. It is converted to fibrin by the action of thrombin in the presence of calcium ions.
Specifications

Specifications

Fibrinogen alpha chain
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
E9PV24, P02671
FGA, Fib2, fibrinogen alpha chain
Rabbit
IgG
150 μL
-20°C
Primary
14161, 2243
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FGA
Liquid
Fga
Fibrinogen alpha chain Fusion Protein Ag9567
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit