Learn More
Alfa Aesar™ Fibrinogen beta-chain (24-42)
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66328MCR
 View more versions of this product
Catalog No. AAJ66328MCR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
1951.14
1mg
Glu-Glu-Ala-Pro-Ser-Leu-Arg-Pro-Ala-Pro-Pro-Pro-Ile-Ser-Gly-Gly-Gly-Tyr-Arg
-30° to -10°C
HPLC
Solid
RUO
Fibrinogen beta-chain (24-42)