Learn More
Thermo Scientific Chemicals Fibrinogen-Binding Inhibitor Peptide
For cellular and molecular biology applications
Supplier: Thermo Scientific Chemicals J60863MCR
Description
Inhibits the binding of fibrinogen and fibronectin
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Solid | |
1189.30 | |
1 mg | |
H-His-His-Leu-Gly-Gly-Ala-Lys-Gln-Ala-Gly-Asp-Val-OH |
Fibrinogen γ-chain dodecapeptide | |
Inhibits the binding of fibrinogen and fibronectin | |
Fibrinogen-Binding Inhibitor Peptide | |
RUO - research use only |