Learn More
Diazyme Laboratories Fibrinogen Control
Diazyme Fibrinogen Control set is for the testing of Fibrinogen in citrated human plasma.
Manufacturer: Diazyme Laboratories DZ768ACON
This item is not returnable.
View return policy
Description
- Highly sensitive liquid stable immunoturbidimetric test kit, designed to work on most open clinical chemistry analyzers to provide rapid results with reduced reagent cost
- The Fibrinogen Assay is linear up to 1032.8 mg/dL with a correlation coefficient value of 0.9973, slope of 0.986 and y-intercept of 2.72
- The following substances do not interfere with this assay at the levels tested less than 10% bias: Hemoglobin up to 1000 mg/dL, Bilirubin 40 mg/dL, Bilirubin conjugated 40mg/dL, Triglycerides 1000 mg/dL, Ascorbic Acid 176 mg/dL, Rheumatoid Factor 220 IU/mL
- For in vitro diagnostic use only
Specifications
510(k) Cleared | |
2°C to 8°C. DO NOT FREEZE. Note: Store CONTROL tightly capped when not in use. | |
Lab dependent | |
The Diazyme Fibrinogen Control Set is a two-level set that is supplied in lyophilized form (2 x 1 mL). The CONTROL set is manufactured from human serum. The concentrations are expressed in mg/dL. |
2 x 1 mL Vial | |
Diagnostic | |
Fibrinogen Assay Control Set | |
Unopened vial: stability is up to the expiration date on label when stored at 2 to 8°C Open vial: stability is 7 days when stored at 2 to 8°C |