Learn More
Fibrinogen gamma chain Antibody (OTI2D2), DyLight 680, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270732FR
Catalog No. NBP270732FR
Add to Cart
Description
Description
Fibrinogen gamma chain Monoclonal specifically detects Fibrinogen gamma chain in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
Fibrinogen gamma chain
Monoclonal
DyLight 680
PBS with 0.05% Sodium Azide
FGG
Full length recombinant protein of human Fibrinogen gamma chain (NP_000500) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Monkey
IgG1
Western Blot, Immunohistochemistry
OTI2D2
Western Blot, Immunohistochemistry
fibrinogen gamma chain, fibrinogen, gamma polypeptide
Mouse
Affinity Purified
Angiogenesis, Cancer
2266.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions