Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrinogen gamma chain Mouse anti-Human, Mouse, Monkey, Alexa Fluor™ 700, Clone: OTI2D2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270732AF700

Catalog No. N270732A700


Add to cart

Description

Description

Fibrinogen gamma chain Monoclonal antibody specifically detects Fibrinogen gamma chain in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Fibrinogen gamma chain
OTI2D2
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
FGG
Full length recombinant protein of human Fibrinogen gamma chain (NP_000500) produced in HEK293T cells.
Immunogen affinity purified
Angiogenesis, Cancer
Primary
2266.0
Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
fibrinogen gamma chain, fibrinogen, gamma polypeptide
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Monkey, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit