Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrinogen gamma chain Antibody (OTI2D2), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270732JF549
Description
Fibrinogen gamma chain Monoclonal specifically detects Fibrinogen gamma chain in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Fibrinogen gamma chain | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FGG | |
Full length recombinant protein of human Fibrinogen gamma chain (NP_000500) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2D2 | |
Western Blot, Immunohistochemistry | |
fibrinogen gamma chain, fibrinogen, gamma polypeptide | |
Mouse | |
Affinity Purified | |
Angiogenesis, Cancer | |
2266.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction