Learn More
Abnova™ Fibrinogen (Human) ELISA Kit
Sandwich enzyme immunoassay for the quantitative measurement of human Fibrinogen
Manufacturer:  Abnova™ KA4794
Catalog No. 89-944-580
Add to cart
Description
Description
  • Used for the in vitro quantitative determination of human fibrinogen in human serum, human plasma, cell lysate and buffered solution
  • Assay will recognize native human fibrinogen
Specifications
Specifications
P02671;P02675;P02679
Human
Biotin
Quantification
AV-HRP
For research use only (RUO)
Store the kit at 4°C.
Buffered Solution, Cell Lysates, Plasma, Serum
15.6 to 1000 ng/mL
Fibrinogen (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human Fibrinogen.
2243
ELISA Kit
100 μL
Colorimetric