Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrinopeptide B, Mouse anti-Human, Clone: 800000, Novus Biologicals™

Mouse Monoclonal Antibody

$135.50 - $419.00

Specifications

Antigen Fibrinopeptide B
Clone 800000
Applications ELISA
Host Species Mouse
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB016304
View Documents
Novus Biologicals
NBP2811100.025MG
0.025 mg
Each for $135.50
Add to cart
 
NB016305
View Documents
Novus Biologicals
NBP2811100.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

Fibrinopeptide B Monoclonal antibody specifically detects Fibrinopeptide B in Human Samples. It is validated for ELISA, In vivo assay.
Specifications

Specifications

Fibrinopeptide B
ELISA
Human
Primary
800000
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit