Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrinopeptide B, Rabbit anti-Human, Clone: 800000, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP2811110.2MG
Description
Fibrin beta-chain Monoclonal specifically detects Fibrin beta-chain in Human samples. It is validated for Flow Cytometry, ELISA, In vivo assay.Specifications
Fibrinopeptide B | |
Monoclonal | |
1 mg/mL | |
ELISA, In vivo assay | |
fibrinogen beta chain, fibrinogen, B beta polypeptide, MGC104327, MGC120405 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
8E5 binds to the amino-terminal region of the beta-chain of human fibrin at an epitope consiting of GHRPLDKC. Binding is possible in both the monomeric form and the polymeric form of the protein. This peptide sequence is found only in fibrin, and not in its precursor fibrinogen and so there is no cross-reactivity with fibrinogen. Fibrin is a non-globular protein which plays a role in the clotting of blood and circulates as the precursor fibrinogen. Polymerization occurs through the action of the protease thrombin, which results in the active fibrin. Fibrin polymerizes as an insoluble matrix and forms a clot along with platelets over a wound site. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG κ |
ELISA | |
800000 | |
Unconjugated | |
PBS | |
FGB | |
8E5 was prepared fusing Sp2/o myeloma cells with spleen cells derived from female BALB/c mice immunized with a synthetic beta-fibrin peptide seqeunce GHRPLDKC conjugated to MB-KLH (maleimidobenzoylated keyhole limpet hemocyanin). | |
0.2 mg | |
Cell Cycle and Replication | |
2244 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction