Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FibrinopeptideA Mouse anti-Human, Clone: D5-B3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266842

Catalog No. NBP266842


Add to cart

Description

Description

FibrinopeptideA Monoclonal antibody specifically detects FibrinopeptideA in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications

Specifications

FibrinopeptideA
D5-B3
Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200
Purified
FGA
Native protein
Protein G purified
Cell Cycle and Replication
Primary
2243
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425
Mouse
IgG1
100μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit