Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibroblast Activation Protein alpha/FAP Rat anti-Mouse, Alexa Fluor 647, Clone: 983802, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB9727R100UG
This item is not returnable.
View return policy
Description
Fibroblast Activation Protein alpha/FAP Monoclonal antibody specifically detects Fibroblast Activation Protein alpha/FAP in Mouse samples. It is validated for Flow CytometrySpecifications
Fibroblast Activation Protein alpha/FAP | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
100 μg | |
Primary | |
Reconstitution default not known | |
Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Flow Cytometry | |
983802 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P97321 | |
Rat | |
RUO | |
2191 | |
Mouse | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title