Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibroblast Antibody (ER-TR7), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10064932B
Description
Fibroblast Monoclonal antibody specifically detects Fibroblast in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)Specifications
Fibroblast | |
Monoclonal | |
Biotin | |
reticular fibroblast | |
Isolated C3H thymic stromal cells | |
0.125 mL | |
Fibroblast Cell Markers | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
ER-TR7 | |
PBS | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction