Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibroblast Growth Factor 18, mouse, Alfa Aesar™

$93.70 - $172.40

Specifications

Molecular Weight 21 kd
Format Lyophilized Powder
Packaging Plastic bottle
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ67127YAM View Documents Alfa Aesar
J67127YAM
5μg Each for $93.70
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ67127EXF View Documents Alfa Aesar
J67127EXF
25μg Each for $172.40
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

21 kd
Lyophilized Powder
Plastic bottle
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit