Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibronectin/Anastellin Mouse anti-Human, HRP, Clone: 2F4, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222113H
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Fibronectin | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
FN1 | |
Purified recombinant fragment of human Fibronectin expressed in E. coli. [UniProt# P02751] | |
0.1 mL | |
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
2335 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
2F4 | |
Western Blot, ELISA, Immunohistochemistry-Paraffin | |
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only