Learn More
Fibronectin, human, stabilized soln., BT-225S, Alfa Aesar™
Used in cell seeding, in cell culture, in murine embroyonic stem cell line (CCE) culture and induction of endothelial cell differentiation. It is also used in extracellular matrix binding assay, in cell migration study and in protein specificity.
Manufacturer:  Alfa Aesar J64738AMP
 View more versions of this product
Catalog No. AAJ64738AMP
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
Liquid
Plastic bottle
10 x 1mg