Learn More
Fibronectin Mouse anti-Human, Bovine, Alexa Fluor™ 532, Clone: A32, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223525AF532
Catalog No. NP2232A32
Add to cart
Description
Description
Fibronectin Monoclonal antibody specifically detects Fibronectin in Human,Bovine samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
Fibronectin
A32
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Clone A32 is highly specific for fibronectin. There is no evidence for cross-reactivity with other connective tissue proteins (vitronectin, elastin, collagen, laminin). Clone A32 cross-reacts with human fibronectin. Other species have not been tested.
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
FN1
Bovine corneal endothelial cells
0.1 mL
Primary
2335
Bovine, Human