Learn More
Fibronectin Mouse anti-Human, Janelia Fluor 646, Clone: A35, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP105056JF646
Catalog No. NBP15056646
Add to cart
Description
Description
Fibronectin Monoclonal antibody specifically detects Fibronectin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
Fibronectin
A35
Immunohistochemistry, Immunohistochemistry-Frozen
Purified
FN1
Lysed bovine corneal endothelial cells and extracellular matrix
Protein A or G purified
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary
2335
Human
Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
There is no evidence for cross-reactivity with other connective tissue protein (vitronectin, elastin, collagen, laminin).