Learn More
Fibronectin Rabbit anti-Mouse, Polyclonal, Biologo
Rabbit Polyclonal Antibody
Manufacturer:  Exalpha Biologicals IncSupplier Diversity Partner FI24951
Encompass
 View more versions of this product
Catalog No. 50-210-0770
Add to cart
Description
Description
Fibronectin Polyclonal antibody specifically detects Fibronectin in Mouse samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays
Specifications
Specifications
Fibronectin
Polyclonal
Unconjugated
FN1
Lyophilized
Rabbit
IgG
0.5 mL
-20°C
Primary
14268
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays
1 mg/mL
PBS with no preservative; pH
P11276
Anastellin; CIG; Cold-insoluble globulin; cumulus cell-specific fibronectin 1 transcript; DKFZp686F10164; DKFZp686H0342; DKFZp686I1370; DKFZp686O13149; E330027I09; ED-B; embryo-specific fibronectin 1 transcript; ferritin L subunit; ferritin L-chain; FIBNEC; Fibronectin; fibronectin 1; fibronectin ED-A- region; fibronectin ED-B+ region; fibronectin variable region; FINC; Fn; FN1; fn-1; FNZ; GFND; GFND2; LETS; LOW QUALITY PROTEIN: fibronectin; migration-stimulating factor; MSF; Ugl-Y1; Ugl-Y2; Ugl-Y3
Purified fibronectin from murine plasma
Affinity chromatography
RUO
Antibody
Polyclonal
Mouse